siRNA Details
| Gene Symbol: SEPT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306147.1 | NP_001293076.1 | 5 | 569 | ||
| NM_018243.3 | NP_060713.1 | 3 | 349 | |||
| XM_006714259.3 | XP_006714322.1 | 3 | 215 | |||
| XM_017008400.1 | XP_016863889.1 | 4 | 358 | |||
| XM_017008401.1 | XP_016863890.1 | 4 | 370 | |||
| XM_017008402.1 | XP_016863891.1 | 4 | 370 | |||
| XM_017008403.1 | XP_016863892.1 | 3 | 310 | |||
| XM_017008404.1 | XP_016863893.1 | 3 | 310 | |||
| GenBank mRNA | AK001711.1 | - | 3 | 252 | ||
| AK302700.1 | - | 5 | 446 | |||
| AL110300.1 | - | 3 | 231 | |||
| BC008083.2 | - | 3 | 335 | 2966709 | ||
| BC063615.1 | - | 3 | 200 | 3848367 | ||
| CR997215.1 | - | 3 | 349 | |||
| DA413789.1 | - | 3 | 223 | |||
| DA497699.1 | - | 3 | 247 | |||
| GU727629.1 | - | 3 | 161 | |||

SEPT11
Genomic Map