siRNA Details
| Gene Symbol: LOC101928659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135180.1 | 1 | 164 | |||
| GenBank mRNA | BQ640484.1 | - | 1 | 161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928659| Gene Symbol: LOC101928659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135180.1 | 1 | 164 | |||
| GenBank mRNA | BQ640484.1 | - | 1 | 161 | ||