siRNA Details
| Gene Symbol: LOC102723471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135668.1 | 1 | 17 | |||
| NR_135669.1 | 1 | 17 | ||||
| NR_135670.1 | 1 | 17 | ||||
| NR_135671.1 | 1 | 17 | ||||
| GenBank mRNA | BX112232.1 | - | 1 | 17 | ||
| BX115740.1 | - | 1 | 14 | |||

LOC102723471
Genomic Map