siRNA Details
| Gene Symbol: LINC01152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110124.1 | 1 | 58 | |||
| NR_110132.1 | 1 | 58 | ||||
| GenBank mRNA | DA197482.1 | - | 1 | 58 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01152| Gene Symbol: LINC01152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110124.1 | 1 | 58 | |||
| NR_110132.1 | 1 | 58 | ||||
| GenBank mRNA | DA197482.1 | - | 1 | 58 | ||