siRNA Details
| Gene Symbol: LOC101928674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110845.1 | 3 | 409 | |||
| NR_110846.1 | 2 | 86 | ||||
| NR_110847.1 | 2 | 353 | ||||
| GenBank mRNA | BG437436.1 | - | 2 | 349 | ||
| HY169606.1 | - | 3 | 409 | |||
| HY173789.1 | - | 2 | 86 | |||

LOC101928674
Genomic Map