siRNA Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278610.1 | NP_001265539.1 | 2 | 366 | ||
| NM_002466.3 | NP_002457.1 | 2 | 366 | |||
| GenBank mRNA | AK223482.1 | - | 2 | 101 | ||
| AK303249.1 | - | 2 | 271 | |||
| AK314791.1 | - | 2 | 271 | |||
| BC007585.1 | - | 2 | 291 | 3162656 | ||
| BC053555.1 | - | 2 | 260 | 6140320 | ||
| BU178833.1 | - | 316 | ||||
| X13293.1 | - | 2 | 228 | |||

MYBL2
Genomic Map