siRNA Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303538.1 | NP_001290467.1 | 3, 4 | 778 | ||
| NM_018273.3 | NP_060743.2 | 4, 5 | 883 | |||
| NR_130317.1 | 2, 3 | 537 | ||||
| GenBank mRNA | AK129801.1 | - | 4, 5 | 586 | ||
| AK297591.1 | - | 2, 3 | 238 | |||
| AK297610.1 | - | 3, 4 | 479 | |||
| BC016919.1 | - | 4, 5 | 575 | 4400601 | ||
| BC040021.1 | - | 4, 5 | 581 | |||

TMEM143
Genomic Map