siRNA Details
| Gene Symbol: C7orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303470.1 | NP_001290399.1 | 3 | 279 | ||
| NM_018275.4 | NP_060745.3 | 3 | 779 | |||
| GenBank mRNA | AK054872.1 | - | 3 | 275 | ||
| AK302342.1 | - | 1 | 47 | |||
| AK310170.1 | - | 4 | 543 | |||
| BC002749.2 | - | 3 | 179 | |||
| BC015722.2 | - | 3 | 737 | 3920412 | ||

C7orf43
Genomic Map