siRNA Details
Gene Symbol: MIATNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110543.1 | 3, 4 | 414 | |||
GenBank mRNA | BQ581362.1 | - | 3, 4 | 430 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIATNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110543.1 | 3, 4 | 414 | |||
GenBank mRNA | BQ581362.1 | - | 3, 4 | 430 |