siRNA Details
| Gene Symbol: LRRC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161575.1 | NP_001155047.1 | 5 | 806 | ||
| NM_018296.5 | NP_060766.5 | 8 | 1076 | |||
| XM_005256025.2 | XP_005256082.1 | 8 | 1076 | |||
| XM_005256027.2 | XP_005256084.1 | 8 | 1076 | |||
| XM_005256028.1 | XP_005256085.1 | 7 | 948 | |||
| XM_011523200.1 | XP_011521502.1 | 8 | 1076 | |||
| XM_011523201.1 | XP_011521503.1 | 5 | 696 | |||
| XM_011523202.2 | XP_011521504.1 | 4 | 551 | |||
| XM_011523204.1 | XP_011521506.1 | 6 | 875 | |||
| XM_011523205.1 | XP_011521507.1 | 7 | 966 | |||
| XM_011523206.2 | XP_011521508.1 | 8 | 1076 | |||
| XM_017023400.1 | XP_016878889.1 | 8 | 1076 | |||
| GenBank mRNA | AK001866.1 | - | 5 | 855 | ||
| AK292311.1 | - | 5 | 785 | |||
| AK301766.1 | - | 5 | 760 | |||
| AY313780.1 | - | 5 | 769 | |||
| AY963267.1 | - | 8 | 1076 | |||
| BC026156.2 | - | 8 | 1076 | 4828487 | ||

LRRC36
Genomic Map