siRNA Details
| Gene Symbol: TRANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014831.2 | NP_055646.2 | 2, 3 | 396 | ||
| XM_006713432.3 | XP_006713495.1 | 3, 4 | 564 | |||
| XM_011534289.2 | XP_011532591.1 | 3, 4 | 1325 | |||
| XM_017007570.1 | XP_016863059.1 | 3, 4 | 367 | |||
| XM_017007571.1 | XP_016863060.1 | 3, 4 | 457 | |||
| XM_017007572.1 | XP_016863061.1 | 3, 4 | 359 | |||
| XM_017007573.1 | XP_016863062.1 | 3, 4 | 367 | |||
| GenBank mRNA | CD698859.1 | - | 2, 3 | 355 | ||
| CN482191.1 | - | 2, 3 | 397 | |||

TRANK1
Genomic Map