siRNA Details
| Gene Symbol: TRANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006713432.3 | XP_006713495.1 | 2 | 364 | ||
| XM_011534289.2 | XP_011532591.1 | 2 | 1125 | |||
| XM_017007570.1 | XP_016863059.1 | 2 | 167 | |||
| XM_017007571.1 | XP_016863060.1 | 2 | 257 | |||
| XM_017007572.1 | XP_016863061.1 | 2 | 159 | |||
| XM_017007573.1 | XP_016863062.1 | 2 | 167 | |||
| GenBank mRNA | CD698859.1 | - | 1 | 155 | ||

TRANK1
Genomic Map