siRNA Details
Gene Symbol: IQCF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145059.1 | NP_001138531.1 | 2 | 316 | ||
GenBank mRNA | BX106781.1 | - | 2 | 316 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IQCF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145059.1 | NP_001138531.1 | 2 | 316 | ||
GenBank mRNA | BX106781.1 | - | 2 | 316 |