siRNA Details
| Gene Symbol: C4orf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104629.1 | NP_001098099.1 | 4 | 368 | ||
| NM_018302.2 | NP_060772.2 | 3 | 363 | |||
| XM_011513712.2 | XP_011512014.1 | 5 | 432 | |||
| XM_011513713.2 | XP_011512015.1 | 4 | 634 | |||
| XM_017008361.1 | XP_016863850.1 | 2 | 1564 | |||
| GenBank mRNA | AK001879.1 | - | 3 | 363 | ||
| BC037906.1 | - | 4 | 329 | 5276648 | ||
| BP220791.1 | - | 3 | 333 | |||
| CF995148.1 | - | 4 | 372 | |||

C4orf19
Genomic Map