siRNA Details
| Gene Symbol: SMIM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145432.1 | NP_001138904.1 | 3 | 552 | ||
| GenBank mRNA | BC006003.1 | - | 3 | 496 | ||
| BC032431.1 | - | 3 | 551 | |||
| BC040154.1 | - | 3 | 457 | |||
| BC046171.2 | - | 3 | 499 | |||
| BC062747.1 | - | 3 | 482 | |||
| BC086307.1 | - | 3 | 515 | |||
| DA733575.1 | - | 3 | 552 | |||

SMIM20
Genomic Map