siRNA Details
| Gene Symbol: CCDC192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317938.1 | NP_001304867.1 | 7 | 722 | ||
| XM_017009805.1 | XP_016865294.1 | 6 | 784 | |||
| XM_017009806.1 | XP_016865295.1 | 7 | 688 | |||
| XM_017009807.1 | XP_016865296.1 | 7 | 679 | |||
| XM_017009808.1 | XP_016865297.1 | 6 | 610 | |||
| XM_017009812.1 | XP_016865301.1 | 6 | 629 | |||

CCDC192
Genomic Map