siRNA Details
Gene Symbol: C5orf66-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105049.1 | 2 | 646 | |||
NR_105050.1 | 2 | 641 | ||||
GenBank mRNA | BE785577.1 | - | 2 | 310 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C5orf66-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105049.1 | 2 | 646 | |||
NR_105050.1 | 2 | 641 | ||||
GenBank mRNA | BE785577.1 | - | 2 | 310 |