siRNA Details
Gene Symbol: LOC101930049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL700839.1 | - | 3 | 270 |
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017009090.1 | XP_016864579.1 | 4 | 1021 | ||
XM_017009092.1 | XP_016864581.1 | 4 | 1021 |
