siRNA Details
| Gene Symbol: LOC101930049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL700839.1 | - | 2, 3 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101930049| Gene Symbol: LOC101930049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL700839.1 | - | 2, 3 | 174 | ||