siRNA Details
| Gene Symbol: LOC100507387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038402.1 | 1, 2 | 145 | |||
| GenBank mRNA | AL117630.1 | - | 1, 2 | 143 | ||
| DA164303.1 | - | 1, 2 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM153A
LOC100507387| Gene Symbol: LOC100507387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038402.1 | 1, 2 | 145 | |||
| GenBank mRNA | AL117630.1 | - | 1, 2 | 143 | ||
| DA164303.1 | - | 1, 2 | 205 | |||