siRNA Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253792.1 | NP_001240721.1 | 5 | 1970 | ||
| NM_018337.3 | NP_060807.2 | 5 | 1973 | |||
| XM_005259034.2 | XP_005259091.1 | 6 | 2078 | |||
| XM_005259035.1 | XP_005259092.1 | 5 | 2010 | |||
| XM_005259036.1 | XP_005259093.1 | 5 | 1877 | |||
| XM_011527068.1 | XP_011525370.1 | 5 | 2036 | |||
| XM_011527069.1 | XP_011525371.1 | 6 | 1928 | |||
| XM_011527070.1 | XP_011525372.1 | 6 | 3622 | |||
| XM_011527071.1 | XP_011525373.1 | 6 | 2049 | |||
| XM_011527073.1 | XP_011525375.1 | 6 | 3619 | |||
| XM_017026932.1 | XP_016882421.1 | 6 | 1911 | |||
| XM_017026933.1 | XP_016882422.1 | 3 | 2222 | |||
| GenBank mRNA | AB052954.1 | - | 5 | 1813 | ||
| AB052955.1 | - | 5 | 1816 | |||
| AF074985.1 | - | 1 | 516 | |||
| AK001999.1 | - | 5 | 1815 | |||
| AK074063.1 | - | 2 | 5528 | |||
| AK097319.1 | - | 2 | 3201 | |||
| AL834130.1 | - | 5 | 1953 | |||
| BC021282.2 | - | 5 | 1800 | 5016494 | ||

ZNF444
Genomic Map