siRNA Details
| Gene Symbol: LOC100129603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126024.1 | 4 | 420 | |||
| GenBank mRNA | BX340435.2 | - | 4 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129603| Gene Symbol: LOC100129603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126024.1 | 4 | 420 | |||
| GenBank mRNA | BX340435.2 | - | 4 | 419 | ||