siRNA Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018342.4 | NP_060812.2 | 5, 6 | 860 | ||
| XM_005263110.1 | XP_005263167.1 | 6, 7 | 815 | |||
| XM_005263112.3 | XP_005263169.1 | 7, 8 | 850 | |||
| XM_006714254.1 | XP_006714317.1 | 6, 7 | 672 | |||
| XM_017008366.1 | XP_016863855.1 | 4, 5 | 828 | |||
| GenBank mRNA | AK002017.1 | - | 4, 5 | 764 | ||
| BC026318.1 | - | 5, 6 | 554 | |||
| BC036551.1 | - | 4, 5 | 2138 | |||
| BC048117.1 | - | 4, 5 | 2133 | |||
| BC054487.1 | - | 5, 6 | 608 | 5732836 | ||
| BC060875.1 | - | 5, 6 | 586 | 4800219 | ||

TMEM144
Genomic Map