siRNA Details
| Gene Symbol: AP5S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204446.1 | NP_001191375.1 | 2, 3 | 531 | ||
| NM_001204447.1 | NP_001191376.1 | 2, 3 | 379 | |||
| NM_018347.2 | NP_060817.1 | 2, 3 | 435 | |||
| GenBank mRNA | AK002030.1 | - | 2, 3 | 403 | ||
| BC043344.1 | - | 2, 3 | 372 | 5505030 | ||
| DA129391.1 | - | 2, 3 | 499 | |||
| DA253969.1 | - | 2, 3 | 435 | |||
| DB190907.1 | - | 2, 3 | 379 | |||

AP5S1
Genomic Map