siRNA Details
| Gene Symbol: LOC105376306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135119.1 | 2 | 350 | |||
| GenBank mRNA | CD674435.1 | - | 2 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376306| Gene Symbol: LOC105376306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135119.1 | 2 | 350 | |||
| GenBank mRNA | CD674435.1 | - | 2 | 349 | ||