siRNA Details
| Gene Symbol: LOC101928381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121570.1 | 5 | 847 | |||
| GenBank mRNA | AK310876.1 | - | 5 | 847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928381| Gene Symbol: LOC101928381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121570.1 | 5 | 847 | |||
| GenBank mRNA | AK310876.1 | - | 5 | 847 | ||