siRNA Details
| Gene Symbol: TRIM77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146162.1 | NP_001139634.1 | 3 | 621 | ||
| GenBank mRNA | JF968445.1 | - | 2 | 168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM77| Gene Symbol: TRIM77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146162.1 | NP_001139634.1 | 3 | 621 | ||
| GenBank mRNA | JF968445.1 | - | 2 | 168 | ||