siRNA Details
| Gene Symbol: C1orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142569.2 | NP_001136041.1 | 10 | 3690 | ||
| NM_018265.3 | NP_060735.3 | 10 | 3626 | |||
| XM_011509754.2 | XP_011508056.1 | 10 | 4405 | |||
| XM_011509755.1 | XP_011508057.1 | 10 | 3667 | |||
| GenBank mRNA | AK001583.1 | - | 3 | 2271 | ||
| AK001763.1 | - | 10 | 3584 | |||
| AK025066.1 | - | 1 | 1445 | |||

C1orf106
Genomic Map