siRNA Details
| Gene Symbol: WDR41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018268.3 | NP_060738.2 | 8 | 994 | ||
| XM_005248552.3 | XP_005248609.1 | 8 | 961 | |||
| XM_011543505.2 | XP_011541807.1 | 8 | 756 | |||
| XM_011543507.2 | XP_011541809.1 | 8 | 961 | |||
| XM_017009606.1 | XP_016865095.1 | 8 | 1404 | |||
| GenBank mRNA | AF115511.1 | - | 8 | 742 | ||
| AK001766.1 | - | 8 | 940 | |||
| AK021855.1 | - | 8 | 780 | |||
| AK023977.1 | - | 8 | 696 | |||
| AK295249.1 | - | 7 | 601 | |||
| AK296423.1 | - | 7 | 658 | |||
| AK300058.1 | - | 6 | 605 | |||
| AK304329.1 | - | 8 | 686 | |||
| AL834138.1 | - | 2 | 1675 | |||
| BC040241.1 | - | 8 | 746 | 3447269 | ||
| KU178690.1 | - | 8 | 674 | |||
| KU178691.1 | - | 8 | 689 | |||

WDR41
Genomic Map