siRNA Details
| Gene Symbol: LOC102723796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023919.1 | XP_016879408.1 | 5 | 627 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723796| Gene Symbol: LOC102723796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023919.1 | XP_016879408.1 | 5 | 627 | ||