siRNA Details
| Gene Symbol: PLA2G2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316722.1 | NP_001303651.1 | 4 | 351 | ||
| XM_017001270.1 | XP_016856759.1 | 2 | 1020 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G2C| Gene Symbol: PLA2G2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316722.1 | NP_001303651.1 | 4 | 351 | ||
| XM_017001270.1 | XP_016856759.1 | 2 | 1020 | |||