siRNA Details
| Gene Symbol: TMEM140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018295.4 | NP_060765.4 | 2 | 1261 | ||
| GenBank mRNA | AK001862.1 | - | 2 | 1203 | ||
| BC020942.1 | - | 2 | 1187 | 4798940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM140| Gene Symbol: TMEM140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018295.4 | NP_060765.4 | 2 | 1261 | ||
| GenBank mRNA | AK001862.1 | - | 2 | 1203 | ||
| BC020942.1 | - | 2 | 1187 | 4798940 | ||