siRNA Details
| Gene Symbol: ZNF83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105549.1 | NP_001099019.1 | 6 | 2478 | ||
| NM_001105550.1 | NP_001099020.1 | 5 | 2362 | |||
| NM_001105551.1 | NP_001099021.1 | 5 | 2388 | |||
| NM_001105552.1 | NP_001099022.1 | 4 | 2264 | |||
| NM_001277945.1 | NP_001264874.1 | 6 | 2532 | |||
| NM_001277946.1 | NP_001264875.1 | 4 | 2317 | |||
| NM_001277947.1 | NP_001264876.1 | 6 | 2477 | |||
| NM_001277948.1 | NP_001264877.1 | 5 | 2405 | |||
| NM_001277949.1 | NP_001264878.1 | 5 | 2444 | |||
| NM_001277951.1 | NP_001264880.1 | 3 | 2188 | |||
| NM_001277952.1 | NP_001264881.1 | 5 | 2382 | |||
| NM_018300.3 | NP_060770.3 | 3 | 2137 | |||
| XM_017026951.1 | XP_016882440.1 | 7 | 2634 | |||
| GenBank mRNA | AB209516.1 | - | 2 | 3277 | ||
| AK001877.1 | - | 2 | 1753 | |||
| AK027518.1 | - | 3 | 2182 | |||
| AK027782.1 | - | 3 | 1918 | |||
| AK055318.1 | - | 2 | 2669 | |||
| AK075066.1 | - | 7 | 2355 | |||
| AK225945.1 | - | 1 | 1888 | |||
| AK289399.1 | - | 5 | 2138 | |||
| AY500361.1 | - | 1 | 935 | |||
| BC032867.1 | - | 7 | 2377 | 5264072 | ||
| BC050407.1 | - | 1 | 1878 | 6197570 | ||
| BC063032.1 | - | 2 | 2026 | 5553802 | ||
| BG260506.1 | - | 395 | ||||
| BX647540.1 | - | 2 | 3994 | |||
| M27877.1 | - | 1 | 1483 | |||

ZNF83
Genomic Map