siRNA Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011532221.2 | XP_011530523.1 | 1 | 72 | ||
| XM_011532222.2 | XP_011530524.1 | 1 | 72 | |||
| XM_011532223.2 | XP_011530525.1 | 1 | 72 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM160A1| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011532221.2 | XP_011530523.1 | 1 | 72 | ||
| XM_011532222.2 | XP_011530524.1 | 1 | 72 | |||
| XM_011532223.2 | XP_011530525.1 | 1 | 72 | |||