siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098176.1 | - | 9 | 1139 | ||
| AK302348.1 | - | 6 | 739 | |||
| AK304443.1 | - | 8 | 900 | |||
| AK316511.1 | - | 8 | 1334 | |||
| Gene Symbol: LOC107984125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014157.1 | XP_016869646.1 | 2 | 348 | ||

RP1
Genomic Map