siRNA Details
Gene Symbol: SAYSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304793.1 | NP_001291722.1 | 3 | 5724 | ||
NM_018322.2 | NP_060792.1 | 2 | 1317 | |||
GenBank mRNA | AK001963.1 | - | 2 | 1220 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SAYSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304793.1 | NP_001291722.1 | 3 | 5724 | ||
NM_018322.2 | NP_060792.1 | 2 | 1317 | |||
GenBank mRNA | AK001963.1 | - | 2 | 1220 |