siRNA Details
| Gene Symbol: GLOD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543912.2 | XP_011542214.1 | 1, 2 | 274 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLOD5| Gene Symbol: GLOD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543912.2 | XP_011542214.1 | 1, 2 | 274 | ||