siRNA Details
| Gene Symbol: COBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011515234.1 | XP_011513536.1 | 6, 7 | 1002 | ||
| XM_011515235.1 | XP_011513537.1 | 5, 6 | 957 | |||
| XM_011515236.1 | XP_011513538.1 | 6, 7 | 1002 | |||
| XM_011515237.1 | XP_011513539.1 | 6, 7 | 1002 | |||
| XM_011515238.1 | XP_011513540.1 | 6, 7 | 1002 | |||
| XM_011515239.1 | XP_011513541.1 | 6, 7 | 1002 | |||
| XM_011515240.1 | XP_011513542.1 | 5, 6 | 957 | |||
| GenBank mRNA | BC045771.1 | - | 2, 3 | 190 | ||
| CR936689.1 | - | 3, 4 | 382 | |||

COBL
Genomic Map