siRNA Details
| Gene Symbol: LOC100507065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120431.1 | 3 | 441 | |||
| NR_120432.1 | 5 | 614 | ||||
| NR_120433.1 | 5 | 618 | ||||
| GenBank mRNA | BE895205.1 | - | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507065| Gene Symbol: LOC100507065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120431.1 | 3 | 441 | |||
| NR_120432.1 | 5 | 614 | ||||
| NR_120433.1 | 5 | 618 | ||||
| GenBank mRNA | BE895205.1 | - | 399 | |||