siRNA Details
| Gene Symbol: LINC00939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137270.1 | - | 4 | 2073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00939| Gene Symbol: LINC00939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137270.1 | - | 4 | 2073 | ||