siRNA Details
| Gene Symbol: LINC00547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040244.1 | 5 | 1903 | |||
| GenBank mRNA | AK092066.1 | - | 5 | 1903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00547| Gene Symbol: LINC00547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040244.1 | 5 | 1903 | |||
| GenBank mRNA | AK092066.1 | - | 5 | 1903 | ||