siRNA Details
| Gene Symbol: TUSC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104174.1 | 2 | 601 | |||
| GenBank mRNA | BC025370.2 | - | 2 | 502 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUSC8| Gene Symbol: TUSC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104174.1 | 2 | 601 | |||
| GenBank mRNA | BC025370.2 | - | 2 | 502 | ||