siRNA Details
| Gene Symbol: LOC101928730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120422.1 | 7 | 1504 | |||
| GenBank mRNA | BC034786.1 | - | 7 | 1503 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928730| Gene Symbol: LOC101928730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120422.1 | 7 | 1504 | |||
| GenBank mRNA | BC034786.1 | - | 7 | 1503 | ||