siRNA Details
| Gene Symbol: PLEKHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161498.1 | NP_001154970.1 | 2 | 414 | ||
| XM_017021290.1 | XP_016876779.1 | 2 | 776 | |||
| GenBank mRNA | AK126770.1 | - | 2 | 476 | ||
| BC146872.1 | - | 2 | 319 | 9056800 | ||

PLEKHD1
Genomic Map