siRNA Details
| Gene Symbol: UFSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018359.3 | NP_060829.2 | 3 | 224 | ||
| NR_028085.1 | 3 | 224 | ||||
| GenBank mRNA | AK002062.1 | - | 3 | 153 | ||
| AK091635.1 | - | 3 | 217 | |||
| BC010493.1 | - | 2 | 120 | 3048997 | ||
| BP264462.1 | - | 3 | 220 | |||
| CR457278.1 | - | 2 | 107 | |||
| DA537391.1 | - | 3 | 218 | |||

UFSP2
Genomic Map