siRNA Details
| Gene Symbol: LOC105370697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135205.1 | 2 | 863 | |||
| GenBank mRNA | BC033241.1 | - | 2 | 863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370697| Gene Symbol: LOC105370697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135205.1 | 2 | 863 | |||
| GenBank mRNA | BC033241.1 | - | 2 | 863 | ||