siRNA Details
| Gene Symbol: RNLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031709.2 | NP_001026879.2 | 3 | 426 | ||
| NM_018363.3 | NP_060833.1 | 3 | 426 | |||
| XM_005269946.1 | XP_005270003.1 | 3 | 352 | |||
| XM_005269947.1 | XP_005270004.1 | 3 | 352 | |||
| XM_005269949.4 | XP_005270006.1 | 3 | 352 | |||
| XM_011539924.2 | XP_011538226.1 | 3 | 352 | |||
| XM_011539927.2 | XP_011538229.1 | 3 | 352 | |||
| XM_017016380.1 | XP_016871869.1 | 3 | 352 | |||
| XM_017016381.1 | XP_016871870.1 | 3 | 352 | |||
| XM_017016383.1 | XP_016871872.1 | 3 | 352 | |||
| GenBank mRNA | AK002080.1 | - | 3 | 353 | ||
| BC005364.1 | - | 3 | 314 | 3828014 | ||
| BP332262.1 | - | 3 | 426 | |||

RNLS
Genomic Map