siRNA Details
| Gene Symbol: RSBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018364.4 | NP_060834.2 | 1 | 291 | ||
| NR_130896.1 | 1 | 291 | ||||
| XM_017001518.1 | XP_016857007.1 | 1 | 291 | |||
| GenBank mRNA | AK002082.1 | - | 1 | 106 | ||
| AK292552.1 | - | 1 | 291 | |||
| BC026155.1 | - | 1 | 113 | 4827655 | ||
| BQ062524.1 | - | 267 | ||||

RSBN1
Genomic Map