siRNA Details
Gene Symbol: CRYM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026675.1 | 3 | 1359 | |||
GenBank mRNA | AK123760.1 | - | 3 | 1360 | ||
BC056679.1 | - | 3 | 1087 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CRYM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026675.1 | 3 | 1359 | |||
GenBank mRNA | AK123760.1 | - | 3 | 1360 | ||
BC056679.1 | - | 3 | 1087 |