siRNA Details
| Gene Symbol: LOC643802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537874.1 | - | 2 | 1938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643802| Gene Symbol: LOC643802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537874.1 | - | 2 | 1938 | ||